Ethyl-4-(hydroxymethyl)piperidine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR904086-1G
APOSOR904086-1GEA 0 EUR
APOSOR904086-1G
Ethyl-4-(hydroxymethyl)piperidine-1-carboxylate 95%
Ethyl-4-(hydroxymethyl)piperidine-1-carboxylate
Formel: C₉H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09752679
CAS-Nummer: 118156-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR