2-(5-Methyl-2-thienyl)azepane 95%

Lieferant: Apollo Scientific

OR904171-1G OR904171-250MG
APOSOR904171-1GEA 1550 EUR
APOSOR904171-1G APOSOR904171-250MG
2-(5-Methyl-2-thienyl)azepane 95%
2-(5-Methyl-2-thienyl)azepane
Formel: C₁₁H₁₇NS
Lagertemperatur: Raumtemperatur
MDL: MFCD05190319
CAS-Nummer: 527674-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR