2-Naphthylisocyanat 95%

Lieferant: Apollo Scientific

OR904280-1G
APOSOR904280-1GEA 162 EUR
APOSOR904280-1G
2-Naphthylisocyanat 95%
2-Naphthylisocyanat

When melted, may contain small amount of sediment.

Formel: C₁₁H₇NO
Siedepunkt: 83 °C
Schmelzpunkt: 52…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014319
CAS-Nummer: 2243-54-1
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR