N1,N1-Diethyl-1,2-benzenediamine 95%

Lieferant: Apollo Scientific

OR904367-1G
APOSOR904367-1GEA 364 EUR
APOSOR904367-1G
N1,N1-Diethyl-1,2-benzenediamine 95%
N1,N1-Diethyl-1,2-benzenediamine
Formel: C₁₀H₁₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01120263
CAS-Nummer: 19056-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR