4-Ethyl-2-methylthiazole-5-carboxylic acid 95%

Lieferant: Apollo Scientific

OR904449-1G OR904449-250MG
APOSOR904449-1GEA 538 EUR
APOSOR904449-1G APOSOR904449-250MG
4-Ethyl-2-methylthiazole-5-carboxylic acid 95%
4-Ethyl-2-methylthiazole-5-carboxylic acid
Formel: C₇H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12027007
CAS-Nummer: 119778-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR