Piperidine-4-carbonitrile hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyanopiperidine hydrochloride

OR904481-25G
APOSOR904481-25GEA 52.2 EUR
APOSOR904481-25G
Piperidine-4-carbonitrile hydrochloride 95+%
Piperidine-4-carbonitrile hydrochloride
Formel: C₆H₁₁ClN₂
Molecular Weight: 146,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037930
CAS-Nummer: 240401-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR