2-Ethynylpyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR904484-1G
APOSOR904484-1GEA
108
EUR
APOSOR904484-1G
2-Ethynylpyrimidine 97%
2-Ethynylpyrimidine
Formel:
C₆H₄N₂ Schmelzpunkt: 95…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698134 CAS-Nummer: 37972-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...