1,2-Diphenylhydrazin 95%

Lieferant: Apollo Scientific

OR904547-100G
APOSOR904547-100GEA 183 EUR
APOSOR904547-100G
1,2-Diphenylhydrazin 95%
1,2-Diphenylhydrazine
Formel: C₁₂H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00003012
CAS-Nummer: 122-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR