2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyrimidine

Lieferant: Apollo Scientific
Achtung

OR904606-1G
APOSOR904606-1GEA 371 EUR
APOSOR904606-1G
2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyrimidine
2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyrimidine
Formel: C₁₂H₇Cl₂N₃
Molecular Weight: 264,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08752799
CAS-Nummer: 143696-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR