2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR904619-1G
APOSOR904619-1GEA 371 EUR
APOSOR904619-1G
2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyridine
2-(2,4-Dichlorophenyl)imidazo[1,2-a]pyridine
Formel: C₁₃H₈Cl₂N₂
Molecular Weight: 263.122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01191735
CAS-Nummer: 63111-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR