o-Anisidiniumchlorid 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methoxyaniline hydrochloride
OR904667-100G
APOSOR904667-100GEA
0
EUR
APOSOR904667-100G
o-Anisidiniumchlorid 98%
o-Anisidiniumchlorid
Formel:
C₇H₁₀ClNO Molecular Weight: 159,6134 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054334 CAS-Nummer: 134-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...