4-Amino-3,5-dimethylphenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3,5-dimethylphenol

OR904670-100G
APOSOR904670-100GEA 365 EUR
APOSOR904670-100G
4-Amino-3,5-dimethylphenol 98%
4-Amino-3,5-xylenol
Formel: C₈H₁₁NO
Molecular Weight: 137.1800 g/mol
Schmelzpunkt: 181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01741459
CAS-Nummer: 3096-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR