2-Cyclopentylpyrrolidine 95%

Lieferant: Apollo Scientific

OR904691-1G
APOSOR904691-1GEA 328 EUR
APOSOR904691-1G
2-Cyclopentylpyrrolidine 95%
2-Cyclopentylpyrrolidine
Formel: C₉H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD02663460
CAS-Nummer: 383127-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR