Lamotrigine

Lieferant: Apollo Scientific

Synonyme: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

OR904704-1G OR904704-25G OR904704-5G
APOSOR904704-1GEA 0 EUR
APOSOR904704-1G APOSOR904704-25G APOSOR904704-5G
Lamotrigine
Lamotrigine
Formel: C₉H₇Cl₂N₅
Molecular Weight: 256.09 g/mol
Schmelzpunkt: 184 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00865333
CAS-Nummer: 84057-84-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR