Lamotrigine
Lieferant: Apollo Scientific
Synonyme:
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine
OR904704-1G
OR904704-25G
OR904704-5G
APOSOR904704-1GEA
0
EUR
APOSOR904704-1G
APOSOR904704-25G
APOSOR904704-5G
Lamotrigine
Lamotrigine
Formel:
C₉H₇Cl₂N₅ Molecular Weight: 256.09 g/mol Schmelzpunkt: 184 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00865333 CAS-Nummer: 84057-84-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...