2-Methyl-1-naphthol 95%

Lieferant: Apollo Scientific

OR904745-1G
APOSOR904745-1GEA 82.8 EUR
APOSOR904745-1G
2-Methyl-1-naphthol 95%
2-Methyl-1-naphthol
Formel: C₁₁H₁₀O
Molecular Weight: 158,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003964
CAS-Nummer: 7469-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR