5-Bromo-1-methyl-1H-1,2,4-triazole 97%

Lieferant: Apollo Scientific

OR904770-5G
APOSOR904770-5GEA 65.4 EUR
APOSOR904770-5G
5-Bromo-1-methyl-1H-1,2,4-triazole 97%
5-Bromo-1-methyl-1H-1,2,4-triazole
Formel: C₃H₄BrN₃
Molecular Weight: 161.9900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109756
CAS-Nummer: 16681-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR