Methyl-5-methylthiazole-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-methylthiazole-2-carboxylate

OR904772-1G
APOSOR904772-1GEA 97.3 EUR
APOSOR904772-1G
Methyl-5-methylthiazole-2-carboxylate 95%
Methyl-5-methylthiazole-2-carboxylate
Formel: C₆H₇NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01924773
CAS-Nummer: 79247-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR