Ethyl 2-methylthiophene-3-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-methylthiophene-3-carboxylate

OR904776-5G
APOSOR904776-5GEA 155 EUR
APOSOR904776-5G
Ethyl 2-methylthiophene-3-carboxylate 98%
Ethyl-2-methylthiophene-3-carboxylate
Formel: C₈H₁₀O₂S
Molecular Weight: 170.2300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698267
CAS-Nummer: 19432-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR