trans-1,4-Cyclohexanedimethanol 97%
Lieferant: Apollo Scientific
Synonyme:
trans-1,4-Bis(hydroxymethyl)cyclohexane
OR904832-25G
APOSOR904832-25GEA
59.6
EUR
APOSOR904832-25G
trans-1,4-Cyclohexanedimethanol 97%
trans-1,4-Cyclohexanedimethanol
Formel:
C₈H₁₆O₂ Molecular Weight: 144.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066360 CAS-Nummer: 3236-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...