2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride 95%
Lieferant: Apollo Scientific
OR905048-1G
APOSOR905048-1GEA
435
EUR
APOSOR905048-1G
2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride 95%
2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride
Formel:
C₈H₁₀ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367043 CAS-Nummer: 173159-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...