5-Bromo-1H-pyrrolo[3,2-b]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-4-azaindole

OR905114-1G
APOSOR905114-1GEA 62.5 EUR
APOSOR905114-1G
5-Bromo-1H-pyrrolo[3,2-b]pyridine 95%
5-Bromo-1H-pyrrolo[3,2-b]pyridine
Formel: C₇H₅BrN₂
Molecular Weight: 197.0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263244
CAS-Nummer: 1000341-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR