tert-Butyl morpholin-2-ylmethylcarbamate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(N-Boc-aminomethyl)morpholine, tert-Butyl (morpholin-2-ylmethyl)carbamate
OR905155-1G
APOSOR905155-1GEA
85.7
EUR
APOSOR905155-1G
tert-Butyl morpholin-2-ylmethylcarbamate 97%
tert-Butyl morpholin-2-ylmethylcarbamate
Formel:
C₁₀H₂₀N₂O₃ Molecular Weight: 216,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808588 CAS-Nummer: 173341-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...