2,4-Dimethylpiperidine hydrochloride 95%

Lieferant: Apollo Scientific

OR905226-1G
APOSOR905226-1GEA 435 EUR
APOSOR905226-1G
2,4-Dimethylpiperidine hydrochloride 95%
2,4-Dimethylpiperidine hydrochloride
Formel: C₇H₁₆ClN
Lagertemperatur: Raumtemperatur
CAS-Nummer: 91846-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR