6-Methoxy-m-toluidin 98%
Lieferant: Apollo Scientific
Synonyme:
1-Amino-2-methoxy-5-methylbenzol, 5-Methyl-o-anisidin, 2-Amino-4-methylanisol, 3-Amino-4-methoxytoluol, p-Cresidin, 2-Methoxy-5-methylanilin
OR905244-100G
APOSOR905244-100GEA
132
EUR
APOSOR905244-100G
6-Methoxy-m-toluidin 98%
6-Methoxy-m-toluidin
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007815 CAS-Nummer: 120-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...