2-(2,4-Dimethylphenyl)pyrrolidine 95%
Lieferant: Apollo Scientific
OR905265-1G
APOSOR905265-1GEA
0
EUR
APOSOR905265-1G
2-(2,4-Dimethylphenyl)pyrrolidine 95%
2-(2,4-Dimethylphenyl)pyrrolidine
Formel:
C₁₂H₁₇N Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663504 CAS-Nummer: 298690-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...