1-(2-Ethylphenyl)-1H-pyrrole 95%

Lieferant: Apollo Scientific

OR905271-1G OR905271-250MG
APOSOR905271-1GEA 458 EUR
APOSOR905271-1G APOSOR905271-250MG
1-(2-Ethylphenyl)-1H-pyrrole 95%
1-(2-Ethylphenyl)-1H-pyrrole
Formel: C₁₂H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD02665229
CAS-Nummer: 299426-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR