4-Methoxyisochinolin 95%

Lieferant: Apollo Scientific

OR905298-1G
APOSOR905298-1GEA 66.7 EUR
APOSOR905298-1G
4-Methoxyisochinolin 95%
4-Methoxyisoquinoline
Formel: C₁₀H₉NO
Molecular Weight: 159,1846 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646265
CAS-Nummer: 36034-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR