5-Methyl-3-(1H-pyrrol-1-yl)-isoxazole 95%
Lieferant: Apollo Scientific
OR905303-1G
APOSOR905303-1GEA
348
EUR
APOSOR905303-1G
5-Methyl-3-(1H-pyrrol-1-yl)-isoxazole 95%
5-Methyl-3-(1H-pyrrol-1-yl)-isoxazole
Formel:
C₈H₈N₂O Molecular Weight: 148,162 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02665522 CAS-Nummer: 358332-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...