2-(3-Methoxyphenyl)pyrrolidine 95%
Lieferant: Apollo Scientific
OR905430-1G
APOSOR905430-1GEA
116
EUR
APOSOR905430-1G
2-(3-Methoxyphenyl)pyrrolidine 95%
2-(3-Methoxyphenyl)pyrrolidine
Formel:
C₁₁H₁₅NO Molecular Weight: 177,243 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663445 CAS-Nummer: 103861-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...