Ethyl-3-amino-3-methylbutanoathydrochlorid 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-amino-3-methylbutanoate hydrochloride

OR905460-25G
APOSOR905460-25GEA 106 EUR
APOSOR905460-25G
Ethyl-3-amino-3-methylbutanoathydrochlorid 95%
Ethyl-3-amino-3-methylbutanoate hydrochloride
Formel: C₇H₁₆ClNO₂
Molecular Weight: 181.6604 g/mol
Schmelzpunkt: 101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11844787
CAS-Nummer: 85532-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR