4-(4-Methylphenyl)-1,3-thiazole 95%
Lieferant: Apollo Scientific
Synonyme:
4-(p-Tolyl)thiazole
OR905477-1G
APOSOR905477-1GEA
317
EUR
APOSOR905477-1G
4-(4-Methylphenyl)-1,3-thiazole 95%
4-(4-Methylphenyl)-1,3-thiazole
Formel:
C₁₀H₉NS Molecular Weight: 175,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02665781 CAS-Nummer: 1826-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...