(2-Ethyl-1H-imidazol-1-yl)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Ethyl-1H-imidazol-1-yl)acetic acid

OR905479-1G
APOSOR905479-1GEA 194 EUR
APOSOR905479-1G
(2-Ethyl-1H-imidazol-1-yl)acetic acid 95%
(2-Ethyl-1H-imidazol-1-yl)acetic acid
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07186426
CAS-Nummer: 883539-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR