2-Amino-2-methyl-butyric acid methyl ester hydrochloride 96%

Lieferant: Apollo Scientific

OR905498-1G
APOSOR905498-1GEA 364 EUR
APOSOR905498-1G
2-Amino-2-methyl-butyric acid methyl ester hydrochloride 96%
2-Amino-2-methyl-butyric acid methyl ester hydrochloride
Formel: C₆H₁₃NO₂·ClH
Molecular Weight: 167,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06737779
CAS-Nummer: 156032-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR