3-Bromo-N,N-dimethylpropan-1-amine hydrobromide 95%

Lieferant: Apollo Scientific

OR905595-5G
APOSOR905595-5GEA 59.6 EUR
APOSOR905595-5G
3-Bromo-N,N-dimethylpropan-1-amine hydrobromide 95%
3-Bromo-N,N-dimethylpropan-1-amine hydrobromide
Formel: C5H13Br2N
Molecular Weight: 246.9714 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11109991
CAS-Nummer: 5845-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR