2-(Aminomethyl)benzonitril Hydrochlorid 96%

Lieferant: Apollo Scientific

OR905627-1G OR905627-5G
APOSOR905627-1GEA 87 EUR
APOSOR905627-1G APOSOR905627-5G
2-(Aminomethyl)benzonitril Hydrochlorid 96%
2-(Aminomethyl)benzonitril Hydrochlorid
Formel: C₈H₉ClN₂
Lagertemperatur: Kühlschrank
MDL: MFCD06797973
CAS-Nummer: 1134529-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR