Methylnicotinat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl-3-pyridincarboxylat, Nicotinsäuremethylester, Pyridin-3-carbonsäuremethylester, 3-Picolinsäuremethylester, 3-Pyridincarbonsäuremethylester

OR905628-500G
APOSOR905628-500GEA 87 EUR
APOSOR905628-500G
Methylnicotinat 98%
Methylnicotinat
Formel: C₇H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00006388
CAS-Nummer: 93-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR