2-Chlor-5-formyl-4-picolin 96%
Lieferant: Apollo Scientific
OR905651-1G
APOSOR905651-1GEA
202
EUR
APOSOR905651-1G
2-Chlor-5-formyl-4-picolin 96%
2-Chlor-5-formyl-4-picolin
Formel:
C₇H₆ClNO Molecular Weight: 155,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368881 CAS-Nummer: 884495-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...