5,7-Dihydroxyimidazo[1,2-a]pyrimidine 95%

Lieferant: Apollo Scientific

OR905656-1G OR905656-250MG
APOSOR905656-1GEA 127 EUR
APOSOR905656-1G APOSOR905656-250MG
5,7-Dihydroxyimidazo[1,2-a]pyrimidine 95%
5,7-Dihydroxyimidazo[1,2-a]pyrimidine
Formel: C₆H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09834820
CAS-Nummer: 51647-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR