6,7-Dimethyl-1H-indole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR905679-1G
APOSOR905679-1GEA 406 EUR
APOSOR905679-1G
6,7-Dimethyl-1H-indole-2-carboxylic acid 95%
6,7-Dimethyl-1H-indole-2-carboxylic acid
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664418
CAS-Nummer: 383132-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR