2-(6-Chloropyrazin-2-yl)acetic acid 95%

Lieferant: Apollo Scientific

OR905690-1G OR905690-250MG
APOSOR905690-1GEA 0 EUR
APOSOR905690-1G APOSOR905690-250MG
2-(6-Chloropyrazin-2-yl)acetic acid 95%
2-(6-Chloropyrazin-2-yl)acetic acid
Formel: C₆H₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09907855
CAS-Nummer: 930798-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR