4-Chlor-6-methyl-2-(methylsulfanyl)pyrimidin  95%

Lieferant: Apollo Scientific

Synonyme: 4-chloro-6-methyl-2-(methylsulphanyl)pyrimidine, 4-Chloro-6-methyl-2-methylthiopyrimidine

OR905698-25G
APOSOR905698-25GEA 126 EUR
APOSOR905698-25G
4-Chlor-6-methyl-2-(methylsulfanyl)pyrimidin  95%
4-Chloro-6-methyl-2-(methylthio)pyrimidine
Formel: C₆H₇ClN₂S
Molecular Weight: 174.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023233
CAS-Nummer: 17119-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR