3-Amino-1-methyl-1H-pyrazole-4-carbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-3-amino-4-cyanopyrazole, 3-amino-1-methyl-1H-pyrazole-4-carbonitrile

OR905717-1G
APOSOR905717-1GEA 451 EUR
APOSOR905717-1G
3-Amino-1-methyl-1H-pyrazole-4-carbonitrile 95%
3-Amino-1-methyl-1H-pyrazole-4-carbonitrile
Formel: C5H6N4
Molecular Weight: 122,131 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 21230-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR