3-[(Methoxymethoxy)methyl]phenylboronic acid 96%
Lieferant: Apollo Scientific
OR905810-1G
OR905810-5G
APOSOR905810-1GEA
183
EUR
APOSOR905810-1G
APOSOR905810-5G
3-[(Methoxymethoxy)methyl]phenylboronic acid 96%
3-[(Methoxymethoxy)methyl]phenylboronic acid
Formel:
C₉H₁₃BO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205753 CAS-Nummer: 2096342-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...