3-Bromo-6-methylpyridazine

Lieferant: Apollo Scientific

OR905842-25G
APOSOR905842-25GEA 482 EUR
APOSOR905842-25G
3-Bromo-6-methylpyridazine
3-Bromo-6-methylpyridazine
Formel: C5H5BrN2
Molecular Weight: 173.0106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09831907
CAS-Nummer: 65202-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR