4,7-Dimethoxy-1,10-phenanthroline 97%

Lieferant: Apollo Scientific

OR905864-5G
APOSOR905864-5GEA 68.3 EUR
APOSOR905864-5G
4,7-Dimethoxy-1,10-phenanthroline 97%
4,7-Dimethoxy-1,10-phenanthroline
Formel: C₁₄H₁₂N₂O₂
Molecular Weight: 240.2573 g/mol
Schmelzpunkt: 197…212 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00233883
CAS-Nummer: 92149-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR