Methyl-3-amino-6-chloropyrazine-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-amino-6-chloropyrazine-2-carboxylate

OR905907-5G
APOSOR905907-5GEA 102 EUR
APOSOR905907-5G
Methyl-3-amino-6-chloropyrazine-2-carboxylate 95%
Methyl-3-amino-6-chloropyrazine-2-carboxylate
Formel: C₆H₆ClN₃O₂
Molecular Weight: 187.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00834965
CAS-Nummer: 1458-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR