5-(tert-Butyl)isoxazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR905958-1G OR905958-5G
APOSOR905958-1GEA 95.7 EUR
APOSOR905958-1G APOSOR905958-5G
5-(tert-Butyl)isoxazole-3-carboxylic acid 95%
5-(tert-Butyl)isoxazole-3-carboxylic acid
Formel: C₈H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07377301
CAS-Nummer: 90607-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR