2-Phenyldihydro-2H-pyran-4(3H)-one 95%
Lieferant: Apollo Scientific
OR905997-1G
APOSOR905997-1GEA
237
EUR
APOSOR905997-1G
2-Phenyldihydro-2H-pyran-4(3H)-one 95%
2-Phenyldihydro-2H-pyran-4(3H)-one
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,2118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11112124 CAS-Nummer: 147688-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...