1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid 98%

Lieferant: Apollo Scientific

OR906017-1G
APOSOR906017-1GEA 140 EUR
APOSOR906017-1G
1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid 98%
1-Methyl-1H-benzo[d]imidazole-6-carboxylic acid
Formel: C₉H₈N₂O₂
Molecular Weight: 176.1720 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11043014
CAS-Nummer: 53484-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR